missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106316
Description
Antibody detects endogenous levels of total MICAL1.
May be a cytoskeletal regulator that connects NEDD9 to intermediate filaments.Identifiants chimiques
| MICAL1 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| MICAL1 | |
| [F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1 | |
| MICAL1 | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TDZ2, Q8VDP3 | |
| 171580, 64780 | |
| A synthesized peptide derived from human MICAL1(Accession Q8TDZ2), corresponding to amino acid residues E437-A487. | |
| Antibody |
Spécifications
| MICAL1 | |
| Polyclonal | |
| Unconjugated | |
| MICAL1 | |
| [F-actin]-monooxygenase MICAL1; CasL interacting molecule MICAL; MICAL; MICAL1; MICAL-1; microtubule associated monooxygenase, calponin and LIM domain containing 1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; NICAL; protein-methionine sulfoxide oxidase MICAL1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171580, 64780 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TDZ2, Q8VDP3 | |
| MICAL1 | |
| A synthesized peptide derived from human MICAL1(Accession Q8TDZ2), corresponding to amino acid residues E437-A487. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |