missing translation for 'onlineSavingsMsg'
Learn More

Mianserin Hydrochloride 98.0+%, TCI America™

Catalog No. M2623200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2623200MG 200 mg
M26231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2623200MG Supplier TCI America Supplier No. M2623200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21535-47-7
Molecular Formula C18H21ClN2
Molecular Weight (g/mol) 300.83
MDL Number MFCD00055072
InChI Key YNPFMWCWRVTGKJ-UHFFFAOYNA-N
Synonym mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
PubChem CID 68551
ChEBI CHEBI:31843
IUPAC Name hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
SMILES [H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 200 mg
Formula Weight 300.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mianserin Hydrochloride
RTECSNumber HP8780000
TSCA No
Recommended Storage Refrigerator