missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MI 2 (MALT1 inhibitor)

Catalog No. 484810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
484810 10 mg
484850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 484810 Supplier Tocris Bioscience™ Supplier No. 4848/10

MALT1 inhibitor

MALT1 inhibitor (IC50 = 5.84 μM). Binds directly to MALT1 and irreversibly suppresses protease function. Decreases NF-κB activity induced by MALT1. Inhibits cell proliferation and MALT1-mediated cleavage activity. Suppresses human TMD8 and HBL-1 activated B cell-like diffuse large B cell lymphoma (ABC-DLBCL) tumor xenografts in mice and primary human ABC-DLBCLs ex vivo.

Chemical Identifiers

CAS 1047953-91-2
Molecular Formula C19H17Cl3N4O3

Specifications

Quantity 10 mg
Formula Weight 455.72
Purity >98%
Molecular Weight (g/mol) 455.72
Chemical Name or Material MI 2 (MALT1 inhibitor)
Recommended Storage Store at +4°C