accessibility menu, dialog, popup

UserName

CAS RN 1047953-91-2

IUPAC Name: 2-chloro-N-{4-[5-(3,4-dichlorophenyl)-3-(2-methoxyethoxy)-1H-1,2,4-triazol-1-yl]phenyl}acetamide
Synonyms: 2-chloro-N-{4-[5-(3,4-dichlorophenyl)-3-(2-methoxyethoxy)-1,2,4-triazol-1-yl]phenyl}acetamide
Molecular Weight (g/mol): 455.72
Molecular Formula: C19H17Cl3N4O3
InChi Key: TWJGQZBSEMDPQP-UHFFFAOYSA-N
SMILES: COCCOC1=NN(C(=N1)C1=CC(Cl)=C(Cl)C=C1)C1=CC=C(NC(=O)CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results