missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MG 132

Numéro de catalogue. AAJ63250MCR
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ63250MCR 1 mg
AAJ63250LB0 5 mg
AAJ63250MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAJ63250MCR Fournisseur Thermo Scientific Chemicals Code fournisseur J63250MCR
Il en reste null

Inhibitor of calpain proteases | CAS: 133407-82-6475.63 g/mol

MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

Solubility
Soluble in ethanol, chloroform, methanol, DMSO.

Notes
Protect from heat. Store away from oxidizing agents.

Identifiants chimiques

CAS 133407-82-6

Spécifications

Color White
Content And Storage Store at -20°C
Format Crystalline Solid
Molecular Weight (g/mol) 475.63
Product Type MG 132
Quantity 1 mg

RUO – Research Use Only