missing translation for 'onlineSavingsMsg'
Learn More

MFGE8 (Lactadherin) Monoclonal Antibody (MFG-06), Invitrogen™

Numéro de catalogue. 01675026
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01675026 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01675026 Fournisseur Thermo Fisher Scientific Code fournisseur MA119053
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

This gene encodes a preproprotein that is proteolytically processed to form multiple protein products. The major encoded protein product, lactadherin, is a membrane glycoprotein that promotes phagocytosis of apoptotic cells. This protein has also been implicated in wound healing, autoimmune disease, and cancer. Lactadherin can be further processed to form a smaller cleavage product, medin, which comprises the major protein component of aortic medial amyloid (AMA). Alternative splicing results in multiple transcript variants.

Spécifications

Antigène MFGE8 (Lactadherin)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MFG-06
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 15mM sodium azide; pH 7.4
Gène MFGE8
Numéro d’acquisition du gène Q08431
Alias du gène AA408458; AGS; AI325141; BA46; breast epithelial antigen BA46; EDIL1; EGF/factor VIII; HMFG; hP47; HsT19888; lactadherin; Lactadherin short form; Medin; Mfge8; MFG-E8; MFGM; MFGME8; MFGMP-E8; milk fat globule glycoprotein; milk fat globule membrane; milk fat globule membrane protein; Milk fat globule-EGF factor 8; milk fat globule-EGF factor 8 protein; MP47; O-acetyl disialoganglioside synthase; O-acetyl GD3 ganglioside synthase; O-acetyltransferase milk fat globule membrane protein; O-acetyltransferase Milk fat glolbule membrane protein; OAcGD3S; P47; SED1; SPAG10; sperm associated antigen 10; sperm surface protein hP47; sperm surface protein SP47
Symboles de gène MFGE8
Espèces hôtes Mouse
Immunogène Human milk fat globule membrane preparation.
Méthode de purification Protein A
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 4240
Espèces cibles Human
Contenu et stockage 4° C, do not freeze
Type de produit Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats