Learn More
Mettler Toledo™ Temperature Calibration Kits
Catalog No. 01909385
Change view
Click to view available options
Without Certificate
Catalog No.
01909385 Without Certificate
1 options
Catalog No. 01909385 Supplier Mettler Toledo™ Supplier No. 00214455
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
  • Adjustment disc with precision thermometer
  • Sample pan handler
Specifications
For Use With (Equipment) Halogen Moisture Analyzer
Includes Adjustment disc with precision thermometer, sample
Type Temperature Calibration Kit