missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ RS-P25 Printer

Catalog No. 01911243
Change view
Click to view available options
Includes:
RS232C, 9 pin male connector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01911243 RS232C, 9 pin male connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01911243 Supplier Mettler Toledo™ Supplier No. 30702967

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides simple printouts on lightfast or adhesive paper.

  • Fast and reliable with a user-friendly operation
  • Supports connected instrument languages
  • Automatically detects settings of connected device
  • Large paper rolls allow printing wiith fewer refills

Specifications

For Use With (Equipment) 2.2 in. Wide Paper
Electrical Requirements 100/240 V 50/60 Hz
Includes RS232C, 9 pin male connector
Type Compact Printer for Standard Paper