missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reference Electrodes

Catalog No. 01911494
Change view
Click to view available options
Junction:
Ceramic
PTFE Sleeve
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Junction
01911494 Ceramic
01911495 PTFE Sleeve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01911494 Supplier METTLER TOLEDO™ Supplier No. 51343190

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A reference electrode to be paired with half-cell measurement electrodes.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • For use with half-cell measuring electrodes

Accessories & Care

  • S7 connector type: Cable S7 to 2 mm Banana (30281923) required
  • InLab Reference: ceramic junction
  • InLab Reference Plus: moveable PTFE sleeve junction
  • InLab Reference Flow: triple ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Specifications

Connector Types S7
Junction Ceramic
Material Glass
Temperature (Metric) Operating 0°C to 100°C
Type Half Cell Reference Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) For routine applications and measurements in Ion-selective half cells
Internal Reference ARGENTHAL
Model InLab™ Reference
Reference Electrolyte 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.72 in.
Length (Metric) 12 cm
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 1.2 cm
Show More Show Less