missing translation for 'onlineSavingsMsg'
Learn More

Mettler-Toledo™ Calibration Substance ME 18872, Caffeine, For the calibration of the thermosystem 900, MilliporeSigma™ Supelco™

Catalog No. 111014743
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014743 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014743 Supplier Merck Emd Millipore Supplier No. 750355GF
Only null left
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

CAS 58-08-2
Physical Form Neat
Linear Formula C8H10N4O2
MDL Number MFCD00005758
Melting Point 234°C to 236.5°C
Synonym Caffeine; 1,3,7-Trimethylxanthine
Type Analytical Standard
Quantity 5 g
Formula Weight 194.19
Grade Analytical Standard