missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole USP, Medisca™

$131.88 - $892.45

Chemical Identifiers

CAS 443-48-1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-025
View Documents
Medisca Pharmaceutique Inc
014604
25 g N/A
Only null left
 
18-048-026
View Documents
Medisca Pharmaceutique Inc
014605
100 g
Each for $131.88
Only null left
 
18-048-027
View Documents
Medisca Pharmaceutique Inc
014608
500 g
Each for $552.03
Only null left
 
18-048-131
View Documents
Medisca Pharmaceutique Inc
014609
1 kg
Each for $892.45
Only null left
 
Description

Description

Metronidazole USP can be white to pale yellow crystals or crystalline powder. It is odorless and is stable in air, but darkens on exposure to light.

Specifications

Chemical Identifiers

443-48-1

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
443-48-1
25 g
USP
Crystalline Powder
White to Light Yellow
99.0 to 101.0%
Videos
SDS
Documents

Documents