Learn More
Metronidazole USP, Medisca™
Supplier: Medisca Pharmaceutique Inc 014604
Description
Metronidazole USP can be white to pale yellow crystals or crystalline powder. It is odorless and is stable in air, but darkens on exposure to light.
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 443-48-1 | |
| 25 g | |
| USP |
| Crystalline Powder | |
| White to Light Yellow | |
| 99.0 to 101.0% |
Chemical Identifiers
| 443-48-1 |