accessibility menu, dialog, popup

UserName

CH3NNHONOO

CAS RN 443-48-1

IUPAC Name: 2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethan-1-ol
Synonyms: metronidazole vertisal elyzol trichopol methronidazole fossyol tricom tricowas B arilin trichex
Molecular Weight (g/mol): 171.16
Molecular Formula: C6H9N3O3
InChi Key: VAOCPAMSLUNLGC-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1CCO)[N+]([O-])=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (5)
  • (3)
Physical Form 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Type 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (6)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)