missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, MilliporeSigma™ Supelco™

Chemical structure: imidazole

$175.16 - $735.96

Specifications

Packaging Poly Bottle
CAS 443-48-1
Physical Form Crystalline
Linear Formula C6H9N3O3
MDL Number MFCD00009750
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012203
View Documents
Merck Emd Millipore
M37615G
5 g
Each for $175.16
Only null left
 
111012201
View Documents
Merck Emd Millipore
M376125G
25 g
Each for $276.93
Only null left
 
111012202
View Documents
Merck Emd Millipore
M3761100G
100 g
Each for $735.96
Only null left
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

Poly Bottle
Crystalline
MFCD00009750
2-Methyl-5-nitroimidazole-1-ethanol
171.15
443-48-1
C6H9N3O3
159°C to 161°C
Analytical Standard
Analytical Standard
Videos
SDS
Documents

Documents