missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metronidazole, MilliporeSigma™ Supelco™
Chemical structure: imidazole
Supplier: Merck Emd Millipore M376125G
Description
Pharmaceutical (small molecule)Specifications
| Poly Bottle | |
| Crystalline | |
| MFCD00009750 | |
| 2-Methyl-5-nitroimidazole-1-ethanol | |
| 25 g | |
| Analytical Standard |
| 443-48-1 | |
| C6H9N3O3 | |
| 159°C to 161°C | |
| Analytical Standard | |
| 171.15 |