missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MW Series Utility Cart with Solid and Wire Shelves
Description
- Stainless steel, chrome shelf and handle material.
- Load capacity: 375 lb. (170.1 kg).
- Fully interchangeable Super Erecta™ shelves.
- Center shelf can be positioned at any height in 1 in. (2.5 cm) increments.
- One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability.
- 18 in. (46 cm) carts have 4 in. diameter (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. diameter (13 cm) casters.
- Three tiers, 375 lbs (170 kg) weight capacity per cart.
- Solid Shelves 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength.
- Raised edges help contain spills and make cleanup easier.
- Sturdy corner castings facilitate mounting.
- Wire Shelves chrome-plated or stainless-steel shelves with trademarked truss design for added strength.
- Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place.
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Specifications
| Product Type | Utility Cart |
| Capacity (Metric) | 170 kg |
| Certifications/Compliance | NSF |
| Color | Silver |
| Description | 3-tier Model |
| Dimensions (L x W x H) | 24 x 18 x 38 in. |
| For Use With (Application) | Transporting Materials |
| Handle Material | Chrome |
| Height (English) | 38 in. |
| Height (Metric) | 97 cm |
| Show More |