missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide hydrochloride, 98%

Catalog No. AC461820050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461820050 5 g
AC461820250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461820050 Supplier Thermo Scientific Chemicals Supplier No. 461820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7232-21-5 | C14H23Cl2N3O2 | 336.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7232-21-5
Molecular Formula C14H23Cl2N3O2
Molecular Weight (g/mol) 336.26
InChI Key RVFUNJWWXKCWNS-UHFFFAOYSA-N
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

Melting Point 171.0°C to 173.0°C
Assay Percent Range 98%
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: soluble in methanol
Formula Weight 336.26
Chemical Name or Material Metoclopramide hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 230-634-5

RUO – Research Use Only