missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfoxide, 99.9% min., OmniSolv, MilliporeSigma™

Catalog No. MMX1456P6
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX1456P6 1 L Poly-coated Glass Bottle
MMX14561 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMX1456P6 Supplier Technidata Supplier No. MX1456P6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Density 1.10g/cm3 at 20°C
Quantity 1 L
Ignition Point 214.85°C (418.7°F)
Packaging Poly-coated Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9% min.
Linear Formula (CH3)2SO
Vapor Pressure 0.053hPa (20°C)
Boiling Point 189°C
Formula Weight 78.1
Chemical Name or Material Methylsulfoxide
Show More Show Less
Recommended Storage Store at +15°C to +25°C