missing translation for 'onlineSavingsMsg'
Learn More

Methylpropyldichlorosilane, 97%, Thermo Scientific™

Catalog No. AC432880250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432880250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432880250 Supplier Thermo Scientific Chemicals Supplier No. 432880250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4518-94-9
Molecular Formula C4H10Cl2Si
Molecular Weight (g/mol) 157.12
InChI Key GNVPGBIHGALKRR-UHFFFAOYSA-N
Synonym methylpropyldichlorosilane, silane, dichloromethylpropyl, dichloromethylpropylsilane, n-propylmethyldichlorosilane, dichloro methyl propylsilane, dichloromethylpropyl-silan, n-propyl methyl dichlorosilane, dichloro methyl propyl silane, dichloro methyl propylsilane gc
PubChem CID 78275
IUPAC Name dichloro-methyl-propylsilane
SMILES CCC[Si](C)(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0300g/mL
Boiling Point 125.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 1.03
Formula Weight 157.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylpropyldichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse m
EINECSNumber 224-843-

RUO – Research Use Only