missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylparaben Sodium, NF, 95-102%, Spectrum™ Chemical
M1277, 5026-62-0, C8H7NaO3
Supplier: Spectrum Chemical Mfg Cor M127712KG
Description
Spectrum™ Chemical Methylparaben Sodium, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 5026-62-0 | |
| Poly Bottle | |
| 12 kg | |
| [Na+].COC(=O)C1=CC=C([O-])C=C1 | |
| 174.13 | |
| NF |
| 100% | |
| C8H7NaO3 | |
| PESXGULMKCKJCC-UHFFFAOYSA-M | |
| sodium 4-(methoxycarbonyl)benzen-1-olate | |
| 95 to 102% | |
| Crystalline Powder or Lumps |
Chemical Identifiers
| 5026-62-0 | |
| 174.13 | |
| sodium 4-(methoxycarbonyl)benzen-1-olate |
| C8H7NaO3 | |
| PESXGULMKCKJCC-UHFFFAOYSA-M | |
| [Na+].COC(=O)C1=CC=C([O-])C=C1 |