missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben Sodium, NF, 95-102%, Spectrum™ Chemical

Catalog No. p-7157741
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604522 125 g Poly Bottle
18604523 500 g Poly Bottle
18604524 2.5 kg Poly Bottle
18604525 12 kg Poly Bottle
18604526 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604522 Supplier Spectrum Chemical Mfg Cor Supplier No. M1277125GM
Only null left

M1277, 5026-62-0, C8H7NaO3

Spectrum™ Chemical Methylparaben Sodium, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.13
InChI Key PESXGULMKCKJCC-UHFFFAOYSA-M
IUPAC Name sodium 4-(methoxycarbonyl)benzen-1-olate
SMILES [Na+].COC(=O)C1=CC=C([O-])C=C1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 95 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps