accessibility menu, dialog, popup

UserName

NaCH3OOO

CAS RN 5026-62-0

IUPAC Name: sodium 4-(methoxycarbonyl)benzen-1-olate
Synonyms: sodium methylparaben
Molecular Weight (g/mol): 174.13
Molecular Formula: C8H7NaO3
InChi Key: PESXGULMKCKJCC-UHFFFAOYSA-M
SMILES: [Na+].COC(=O)C1=CC=C([O-])C=C1
Percent Purity 
  • (5)
  • (1)
Physical Form 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)