missing translation for 'onlineSavingsMsg'
Learn More

Methylmaleic Hydrazide 98.0+%, TCI America™

Catalog No. M21825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21825G 5 g
M218225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21825G Supplier TCI America Supplier No. M21825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5754-18-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00067746,MFCD00023867
InChI Key QAVYOWFNXMHVEL-UHFFFAOYSA-N
Synonym 4-methylpyridazine-3,6-diol, 3,6-dihydroxy-4-methylpyridazine, 1,2-dihydro-4-methyl-3,6-pyridazinedione, 4-methyl-3,6-pyridazinediol, 3,6-pyridazinedione, 1,2-dihydro-4-methyl, 3, 1,2-dihydro-4-methyl, 2-butenedioic acid, cyclic hydrazide, z, acmc-1aqja, 4-methylmaleic hydrazide, methylmaleic acid hydrazide
PubChem CID 79826
IUPAC Name 4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
SMILES CC1=CC(=O)NNC1=O

Specifications

Melting Point 287°C
Color White-Yellow
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methylmaleic Hydrazide