missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylmagnesium chloride, 3M (22 wt.%) solution in THF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Boiling Point | 66.0°C |
| Linear Formula | CH3MgCl |
| Density | 1.0100g/mL |
| Solubility Information | Solubility in water: reacts. |
| Formula Weight | 74.79 |
| Percent Purity | 22 to 24% |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 1.01 |
| Quantity | 800 mL |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. May cause drowsiness or dizziness. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
| EINECSNumber | 211-629-7 |
| TSCA | TSCA |
RUO – Research Use Only