missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylhydroquinone 98.0+%, TCI America™
Supplier: TCI America M0342100G
Specifications
| Methylhydroquinone | |
| 126°C | |
| MFCD00002345 | |
| methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol | |
| CC1=CC(O)=CC=C1O | |
| 124.14 | |
| 124.14 | |
| Crystalline Powder |
| 95-71-6 | |
| C7H8O2 | |
| 100 g | |
| CNHDIAIOKMXOLK-UHFFFAOYSA-N | |
| 2-methylbenzene-1,4-diol | |
| 7253 | |
| ≥98.0% (T) |
Chemical Identifiers
| 95-71-6 | |
| 124.14 | |
| CNHDIAIOKMXOLK-UHFFFAOYSA-N | |
| 7253 | |
| CC1=CC(O)=CC=C1O |
| C7H8O2 | |
| MFCD00002345 | |
| methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol | |
| 2-methylbenzene-1,4-diol |
Safety and Handling
EINECSNumber : (3)-0546&(4)-0057
RTECSNumber : MX6700000
TSCA : Yes