missing translation for 'onlineSavingsMsg'
Learn More

Methylhydroquinone 98.0+%, TCI America™

Catalog No. p-7132110
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M034225G 25 g
M0342100G 100 g
M0342500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M034225G Supplier TCI America Supplier No. M034225G
Only null left

Chemical Identifiers

CAS 95-71-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002345
InChI Key CNHDIAIOKMXOLK-UHFFFAOYSA-N
Synonym methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol
PubChem CID 7253
IUPAC Name 2-methylbenzene-1,4-diol
SMILES CC1=CC(O)=CC=C1O

Specifications

Melting Point 126°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylhydroquinone
EINECSNumber (3)-0546&(4)-0057
RTECSNumber MX6700000
TSCA Yes