accessibility menu, dialog, popup

UserName

CAS RN 95-71-6

IUPAC Name: 2-methylbenzene-1,4-diol
Synonyms: 1,4-benzenediol, 2-methyl-
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: CNHDIAIOKMXOLK-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1O
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (6)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)