missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Methylhydroquinone
Description
Methylhydroquinone
Spécifications
Spécifications
| CAS | 95-71-6 |
| Melting Point | 128°C to 130°C (lit.) |
| Molecular Formula | C7H8O2 |
| Linear Formula | CH3C6H3-1,4-(OH)2 |
| MDL Number | MFCD00002345 |
| Quantity | 1 kg |
| Synonym | Toluhydroquinone |
| Molecular Weight (g/mol) | 124.14 |
| Percent Purity | 99% |