missing translation for 'onlineSavingsMsg'
Learn More

Methylguanidine hydrochloride, 98%

Catalog No. p-7041725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6003306 5 g
AAJ6003314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6003306 Supplier Thermo Scientific Chemicals Supplier No. J6003306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22661-87-6 | C2H8ClN3 | 109.557 g/mol

Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

Solubility
Soluble in water (50 mg/ml - clear, colorless to light yellow solution).

Notes
Hygroscopic, store away form water/moisture. Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.

Chemical Identifiers

CAS 22661-87-6
Molecular Formula C2H8ClN3
Molecular Weight (g/mol) 109.557
MDL Number MFCD00012576
InChI Key VJQCNCOGZPSOQZ-UHFFFAOYSA-N
Synonym 1-methylguanidine hydrochloride, methylguanidine hydrochloride, methylguanidine hcl, n-methylguanidine hydrochloride, 1-methylguanidine hcl, guanidine, methyl-, hydrochloride, 2-methylguanidine hydrochloride, 1-methylguanidine hydrochloride 1:x, guanidine, methyl-, monohydrochloride, methyl guanidine hcl
PubChem CID 146724
IUPAC Name 2-methylguanidine;hydrochloride
SMILES CN=C(N)N.Cl

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml- clear,colorless to light yellow solution).
Formula Weight 109.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methylguanidine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 245-147-3
RTECSNumber MF3990000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only