missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, Technical, Spectrum™ Chemical

Catalog No. 18611796
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611796 500 mL Amber Glass Bottle
18611797 1 L Amber Glass Bottle
18611798 4 L Amber Glass Bottle
18611794 20 L Steel Can
18611795 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611796 Supplier Spectrum Chemical Mfg Cor Supplier No. M1252500MLGL

Chemical Identifiers

CAS 75-09-2
Molecular Weight (g/mol) 84.93

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Boiling Point 39.8°C
Grade Technical
Chemical Name or Material Methylene Chloride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.