missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, NF, 99%, Spectrum™ Chemical

Numéro de catalogue. p-7157738
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18604513 100 mL Amber Glass Bottle
18604514 500 mL Amber Glass Bottle
18604516 1 L Amber Glass Bottle
18604515 6 x 500 mL Amber Glass Bottle
18604517 4 L Amber Glass Bottle
18604518 6 x 1 L Amber Glass Bottle
18604519 4 x 4 L Amber Glass Bottle
18604520 20 L Steel Can
18604521 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Numéro de catalogue. 18604513 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur M1266100ML
Il en reste null

M1266, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, NF is used as a carrier for pharmaceutical tablet coatings. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Identifiants chimiques

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Spécifications

CAS Max % 100%
Quantity 100 mL
Packaging Amber Glass Bottle
UN Number UN1593
Boiling Point 39.8°C
Percent Purity 99%
Grade NF
Chemical Name or Material Methylene Chloride