missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue 98.0+%, TCI America™

Catalog No. A51051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51051G Supplier TCI America Supplier No. A51051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61-73-4
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.851
MDL Number MFCD00012111
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 319.85
Percent Purity ≥98.0% (T)
Chemical Name or Material Methylene Blue [Ion association reagent for spectrophotometric analysis]
EINECSNumber (5)-1995
RTECSNumber SO5600000
TSCA Yes