missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue hydrate, pure

Catalog No. AC414255000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414255000 500 g Glass bottle
AC414251000 100 g Glass bottle
AC414250025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414255000 Supplier Thermo Scientific Chemicals Supplier No. 414255000
Only null left

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

Melting Point 100.0°C to 110.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Infrared Spectrum Authentic
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
Formula Weight 319.85
Loss on Drying 22% max.
Percent Purity ≥70% (TiCl3-titration)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 500 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methylene Blue hydrate
Show More Show Less
Hazard Category Warning

RUO – Research Use Only