missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Spécifications
Spécifications
| Melting Point | 100.0°C to 110.0°C |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol |
| Infrared Spectrum | Authentic |
| IUPAC Name | [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate |
| Formula Weight | 319.85 |
| Loss on Drying | 22% max. |
| Percent Purity | ≥70% (TiCl3-titration) |
| Grade | Pure |
| Packaging | Glass bottle |
| Color | Green |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Warning |
RUO – Research Use Only