missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 1% (w/v) Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M2041LT

 View more versions of this product

Catalog No. 18611765


Only null left

Description

Description

Specifications

Chemical Identifiers

61-73-4 , 7732-18-5 , 7220-79-3
Chemical Solution
Blue
1%
Poly Bottle
Methylene Blue

Specifications

Methylene Blue
1%
Poly Bottle
1 L
61-73-4 , 7732-18-5 , 7220-79-3
Chemical Solution
Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.