missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 1% (w/v) Aqueous Solution, Spectrum™ Chemical

Catalog No. p-7165895
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18611764 125 mL
18611766 500 mL
18611765 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611764 Supplier Spectrum Chemical Mfg Cor Supplier No. M204125ML
Only null left

Specifications

Chemical Name or Material Methylene Blue
CAS 61-73-4, 7732-18-5, 7220-79-3
Concentration 1%
Grade Chemical Solution
Packaging Poly Bottle
Color Blue
Quantity 125 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.