missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 0.1% Aqueous, Certified, LabChem™

Catalog No. LC169002
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC169002 1 L Poly Bottle
LC169001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC169002 Supplier LabChem Supplier No. LC169002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61-73-4 , 7732-18-5
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.851
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Absorbance Passes Test
CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C16H18N3SCl
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Color Blue
Solubility Information Soluble in water
Quantity 1 L
Physical Form Liquid
Formula Weight 319.85
Grade Certified
Chemical Name or Material Methylene Blue, 0.1% Aqueous
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature