missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-chloro-3,4-dimethoxybenzoate, 97%, Thermo Scientific™

Numéro de catalogue. BTB09269EA
Modifier l'affichage
Click to view available options
Quantité:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
BTB09269EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. BTB09269EA Fournisseur Thermo Scientific Chemicals Code fournisseur BTB09269EA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 175136-01-3
Formule moléculaire C10H11ClO4
Poids moléculaire (g/mol) 230.644
Numéro MDL MFCD00085114
Clé InChI RYYAHUFRPCBHCT-UHFFFAOYSA-N
Synonyme maybridge1_001981, benzoic acid,2-chloro-3,4-dimethoxy-, methyl ester
PubChem CID 2774981
Nom de l’IUPAC methyl 2-chloro-3,4-dimethoxybenzoate
SOURIRES COC1=C(C(=C(C=C1)C(=O)OC)Cl)OC

Spécifications

Nom ou substance chimique Methyle2-chloro-3, 4-dimethoxybenzoate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 10g
Poids de formule 230.65
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.