missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylsulfonium tetrafluoroborate, 95%

Catalog No. AAL0051214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0051214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0051214 Supplier Thermo Scientific Chemicals Supplier No. L0051214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10504-60-6 | C13H13BF4S | 288.11 g/mol

Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

Solubility
Soluble in water, alcohol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 10504-60-6
Molecular Formula C13H13BF4S
Molecular Weight (g/mol) 288.11
MDL Number MFCD00051861
InChI Key LHAMVDBAOJCBDW-UHFFFAOYSA-N
Synonym diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
PubChem CID 2758717
IUPAC Name methyl(diphenyl)sulfanium;tetrafluoroborate
SMILES F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 63°C to 67°C
Linear Formula (C6H5)2SCH3BF4
Quantity 25 g
UN Number UN1759
Beilstein 4732019
Solubility Information Soluble in water,alcohol.
Formula Weight 288.11
Percent Purity 95%
Chemical Name or Material Methyldiphenylsulfonium tetrafluoroborate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 234-032-3
TSCA No

RUO – Research Use Only