missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyldibromoglutaronitrile, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 35691-65-7 |
| Physical Form | Neat |
| Linear Formula | C6H6 Br2N2 |
| MDL Number | MFCD00072483 |
| Melting Point | 51°C to 55°C |
| Synonym | 1,2-Dibromo-2,4-dicyanobutane; 1-Bromo-1-(bromomethyl)-1,3-propanedicarbonitrile; 2-Bromo-2-(bromomethyl)glutaronitrile |
| Type | Analytical Standard |
| Quantity | 500 mg |
| Formula Weight | 265.93 |
| Grade | Analytical Standard |