missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet, 0.02% Aqueous, pH 0.0 to 1.4 Green to Blue, Certified, LabChem™

Catalog No. LC172001
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC172001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC172001 Supplier LabChem Supplier No. LC172001

Chemical Identifiers

CAS 8004-87-3 , 7732-18-5

Specifications

CAS Max % 99.98, 0.02
Linear Formula C24H28N3Cl
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 393.96
Grade Certified
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Purple
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Methyl Violet, 0.02% Aqueous, pH 0.0 to 1.4 Green to Blue
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature