missing translation for 'onlineSavingsMsg'
Learn More

Methyl vanillate, 99%, Thermo Scientific™

Numéro de catalogue. AC160301000
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
100g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC160301000 100g Glass bottle
AC160300050 5g Glass bottle
AC160300250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC160301000 Fournisseur Thermo Scientific Chemicals Code fournisseur 160301000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 3943-74-6
Formule moléculaire C9H10O4
Poids moléculaire (g/mol) 182.18
Numéro MDL MFCD00008438
Clé InChI BVWTXUYLKBHMOX-UHFFFAOYSA-N
Synonyme methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
PubChem CID 19844
ChEBI CHEBI:46477
Nom de l’IUPAC methyl 4-hydroxy-3-methoxybenzoate
SOURIRES COC1=C(C=CC(=C1)C(=O)OC)O

Spécifications

Nom ou substance chimique Methyl vanillate
Point d’ébullition 285.0°C to 287.0°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98.5% min. (GC)
Emballage Glass bottle
Formule linéaire HOC6H3(OCH3)CO2CH3
Quantité 100g
Beilstein 10, 396
Poids de formule 182.18
Pourcentage de pureté 99%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 223-525-9

RUO – Research Use Only