missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Valerate 99.0+%, TCI America™
Chemical Identifiers
| CAS | 624-24-8 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00009478 |
| InChI Key | HNBDRPTVWVGKBR-UHFFFAOYSA-N |
| Synonym | methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn |
| PubChem CID | 12206 |
| IUPAC Name | methyl pentanoate |
| SMILES | CCCCC(=O)OC |
Chemical Identifiers
| C6H12O2 | |
| MFCD00009478 | |
| methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn | |
| methyl pentanoate |
Specifications
| 624-24-8 | |
| 128°C | |
| MFCD00009478 | |
| methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn | |
| CCCCC(=O)OC | |
| 116.16 | |
| 116.16 | |
| Liquid |
| Colorless | |
| C6H12O2 | |
| 3272 | |
| HNBDRPTVWVGKBR-UHFFFAOYSA-N | |
| methyl pentanoate | |
| 12206 | |
| ≥99.0% (GC) | |
| Methyl Valerate |
Safety and Handling
EINECSNumber : (2)-0776
RTECSNumber : YV7750500
TSCA : Yes