missing translation for 'onlineSavingsMsg'
Learn More

Methyl undecanoate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 426580250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC426580250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1731-86-8
200.32
XPQPWPZFBULGKT-UHFFFAOYSA-N
15607
methyl undecanoate
C12H24O2
MFCD00008957
undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
CHEBI:87527
CCCCCCCCCCC(=O)OC

Specifications

Methyl undecanoate
0.8720g/mL
109°C
96% min. (GC)
C12H24O2
CH3(CH2)9CO2CH3
MFCD00008957
undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
XPQPWPZFBULGKT-UHFFFAOYSA-N
methyl undecanoate
15607
200.32
1731-86-8
246°C to 248°C
Authentic
Glass bottle
1.429
25g
0.872
Solubility in water: insoluble
CCCCCCCCCCC(=O)OC
200.32
CHEBI:87527
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-053-2

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...