missing translation for 'onlineSavingsMsg'
Learn More

Methyl trimethylacetate, 99%

Catalog No. AAA1376436
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1376436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1376436 Supplier Thermo Scientific Chemicals Supplier No. A1376436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 598-98-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-98-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008843
InChI Key CNMFHDIDIMZHKY-UHFFFAOYSA-N
Synonym methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
PubChem CID 69027
IUPAC Name methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC

Specifications

Melting Point <-70°C
Density 0.875
Boiling Point 100°C to 101°C
Flash Point 6°C (42°F)
Refractive Index 1.39
Quantity 500 g
UN Number UN3272
Beilstein 1744141
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Methyl trimethylacetate
Show More Show Less
Hazard Category H225-H302
Hazard Statement GHS H Statement
H225-H301
Highly flammable liquid and vapour.
Toxic if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 209-959-1
RTECSNumber UA2459597
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only