missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoromethanesulfonate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 415011000

Catalog No. AC415011000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Alkylation reagent
Specifications

Specifications

Methyl trifluoromethanesulfonate
1.4500g/mL
38°C
95% min. (GC)
C2H3F3O3S
CF3SO3CH3
100g
1.45
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
COS(=O)(=O)C(F)(F)F
164.10
164.11
333-27-7
95.0°C
Authentic
Glass bottle
1.3250 to 1.3270
MFCD00000409
06,406; 12,329; 13,327; 17,191
15,6204
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
methyl trifluoromethanesulfonate
9526
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-371-7

SDS
Documents

Documents

Promotions

Promotions