missing translation for 'onlineSavingsMsg'
Learn More

Méthyl trans-p-coumarate 98,0+%, TCI America™

Catalog No. M22595G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
M22595G 5 g
M225925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22595G Supplier TCI America Supplier No. M22595G

Chemical Identifiers

CAS 19367-38-5
Formule moléculaire C10H10O3
Poids moléculaire (g/mol) 178.19
Numéro MDL MFCD00157167
Clé InChI NITWSHWHQAQBAW-UHFFFAOYSA-N
Synonyme trans-p-Coumaric Acid Methyl Ester, trans-4-Hydroxycinnamic Acid Methyl Ester, Methyl trans-4-Hydroxycinnamate
PubChem CID 5319562
ChEBI CHEBI:86904
Nom de l’IUPAC méthyle 3-(4-hydroxyphényl)prop-2-énoate
SOURIRES COC(=O)C=CC1=CC=C(O)C=C1

Specifications

Nom ou substance chimique Methyl trans-p-Coumarate
Point de fusion 139 °C
Quantité 5 g
Poids de formule 178.19
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline