missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tiglate 98.0+%, TCI America™

Catalog No. T024825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T024825ML 25 mL
T0248100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T024825ML Supplier TCI America Supplier No. T024825ML

Chemical Identifiers

CAS 6622-76-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00016654
InChI Key YYJWBYNQJLBIGS-PLNGDYQASA-N
Synonym methyl tiglate, tiglic acid methyl ester, methyl 2-methylbut-2-enoate, methyl 2-methyl-2-butenoate, methyl e-2-methylcrotonate, methyl 2-methylcrotonate, methyl alpha-methylcrotonate, methyl trans-2-methylcrotonate, 2-butenoic acid, 2-methyl-, methyl ester, e, crotonic acid, 2-methyl-, methyl ester, e
PubChem CID 5323652
IUPAC Name methyl (2Z)-2-methylbut-2-enoate
SMILES COC(=O)C(\C)=C/C

Specifications

Color Colorless
Boiling Point 139°C
Quantity 25 mL
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Tiglate
EINECSNumber (2)-0969
RTECSNumber EM9254600
TSCA Yes