missing translation for 'onlineSavingsMsg'
Learn More

Méthyl téréphtalaldéhydate 98,0+%, TCI America™

Catalog No. T0012500G
Change view
Click to view available options
Quantité:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
T0012500G 500 g
T001225G 25 g
T0012100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0012500G Supplier TCI America Supplier No. T0012500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1571-08-0
Formule moléculaire C9H8O3
Poids moléculaire (g/mol) 164.16
Numéro MDL MFCD00006950
Clé InChI FEIOASZZURHTHB-UHFFFAOYSA-N
Synonyme methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
PubChem CID 15294
Nom de l’IUPAC méthyle 4-formylbenzoate
SOURIRES COC(=O)C1=CC=C(C=O)C=C1

Specifications

Nom ou substance chimique Methyl Terephthalaldehydate
Point de fusion 60 °C
Couleur Blanc
Point d’ébullition 135 °C
Quantité 500 g
Poids de formule 164.16
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline
EINECSNumber (3)-1661
TSCA Oui