missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (S)-(+)-3-hydroxybutyrate, 99%, Thermo Scientific™
Specifications
Specifications
| Density | 1.0550g/mL |
| Boiling Point | 63°C (10.0mmHg) |
| Flash Point | 73°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4190 to 1.422 |
| Quantity | 1g |
| Specific Gravity | 1.055 |
| Specific Rotation Condition | +21.5° (20°C c=neat) |
| Show More |
Safety and Handling
| EINECSNumber | 258-628-8 |
RUO – Research Use Only